Learn More
BRAND™ Polypropylene Cap Inserts
Product Code. 10771791 Shop All Brand Products
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 500
Product Code. 10771791
Brand: BRAND™ 114852
Only null left
Add to basket
Add to basket
Providing a secure fit for all sizes.
  • Available in different colors
  • Provided in packs of 500
  • High temperature and chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Material Polypropylene
Quantity 500/Pk.