Learn More
Kartell™ Polypropylene Conical Erlenmeyer Flasks
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
Brand: Kartell™ 0146500
58.32 EUR valid until 2024-05-31
Use promo code "21660" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
- For general laboratory use
- Provided with standardized neck
- Autoclavable
- Suitable for foodstuff
Specifications
Translucent | |
Polypropylene | |
34/35 NS | |
Conical | |
Yes |
2000 mL | |
100 mL | |
4 | |
Autoclavable |