Learn More
Buerkle™ Polypropylene Container Connector
Product Code. p-8003073 Shop All Bürkle Products
Click to view available options
Width (Metric):
18 mm
24 mm
Color:
Gray
White
Unit Size:
Each
Product Code. 10360391
Brand: Buerkle™ 85300034
Only null left
Add to basket
Add to basket
Container connectors with outer thread for the easy mounting of thread in the sides of containers.
  • With outer thread
  • Complete with EPDM seals
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs
Description Outer thread
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Used for Easy Mounting of Thread in The Sides of Containers
Thread Size 3/4 in.
Width (Metric) 18 mm
Includes 3/4 in. Thread size
Color Gray
Product Type Container Connection