Learn More
Huenersdorf™ Specimen Containers
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 14
Pack of 25
Description
- Resistant to acids and chemicals
- With tightly closing lid
- Deep freezable
Specifications
Specifications
| Product Type | Container |
| Capacity (Metric) | 500 mL |
| Diameter (Metric) | 104 mm |
| Height (Metric) | 100 mm |
| No. per Pack | 14/Pk. |