missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Polypropylene Cryoboxes

Product Code. 12479350 Shop All TENAK Products
Click to view available options
Color:
Blue
Neutral
Red
Yellow
Holds:
1.2 to 2 mL Cryogenic Vials
Centrifuge Tubes
Unit Size:
Pack of 36
This item is not returnable. View return policy

Product Code. 12479350

Brand: Tenak™ TE22292

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Color Neutral
Material Polypropylene
Shape Square
Type Cryo Box
Holds Centrifuge Tubes
Quantity 36 Pack
Width (Metric) 130 mm
Height (Metric) 50 mm
Weight (Metric) 0.08 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.