Learn More
Simport™ Polypropylene Cryovials
Designed for storing biological material, human or animal cells at temperatures as low as -196 °C. Simport™ Polypropylene Cryovials ensure an equally secure seal both at room temperature and at low cryogenic temperatures.
Brand:  Simport™ T311-2
Additional Details : Weight : 0.80000kg
Product Code. 10407362
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Capinserts are available for color-coding
  • Vertical ribs facilitate cap removal
  • Silicone washer
  • Super-fast 1 1/4 turn thread design
  • Both cap and tube are made of same polypropylene material, therefore same coefficient of expansion
  • ensures secure seal at all temperatures
  • Thick wall makes vial almost unbreakable
  • Large white marking area
  • Excellent clarity makes sample easy to see
  • Round bottom
  • Very easy to empty contents completely