Learn More
Simport™ Polypropylene Cryovials
Designed for storing biological material, human or animal cells at temperatures as low as -196 °C. Simport™ Polypropylene Cryovials ensure an equally secure seal both at room temperature and at low cryogenic temperatures.
Brand: Simport™ T311-2
Additional Details : Weight : 0.80000kg
Description
- Capinserts are available for color-coding
- Vertical ribs facilitate cap removal
- Silicone washer
- Super-fast 1 1/4 turn thread design
- Both cap and tube are made of same polypropylene material, therefore same coefficient of expansion
- ensures secure seal at all temperatures
- Thick wall makes vial almost unbreakable
- Large white marking area
- Excellent clarity makes sample easy to see
- Round bottom
- Very easy to empty contents completely