Learn More
Brand™ Polypropylene Wide Neck Erlenmeyer Flasks
Product Code. 10799581 Shop All Brand Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
1000 mL
Unit Size:
Pack of 4
Pack of 6
Product Code. 10799581
Brand: Brand™ 93172
Only null left
Add to basket
Add to basket
Flasks with high clarity for visibility of contents during routine laboratory procedures.
  • Provided with screw cap closure
  • With blue graduation marks
  • Neck with standard ground socket
  • Provided in packs of 6, except for 1000mL flask, which comes in a pack of 4
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Material Polypropylene
Height (Metric) 110 mm
Neck Style 34/35
No. per Pack 6
Thread Size GL 40
Closure Type Screw Cap
Diameter (Metric) Outer 64 mm