Learn More
Cytiva Polypropylene Flangless Connector
Product Code. 11799565 Shop All Cytiva Products
Change view
Click to view available options
Connections:
TC-2 Female to Female
Unit Size:
Pack of 2
Product Code. Connections unitSize
11799565 TC-2 Female to Female Pack of 2
1 options
Product Code. 11799565
Brand: Cytiva 19004501
Only null left
Specifications
Connections TC-2 Female to Female
Temperature (Metric) Max 60°C
For Use With (Equipment) 1.8 mm O.D. Tubing