Learn More
Huenersdorf™ Polypropylene Graduated Beaker with Closed Handle and 2 Scales
Product Code. 12055605
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Product Code. 12055605
Brand: Huenersdorf™ 935000
Only null left
Add to basket
Edge
Add to basket
Huenersdorf™ Polypropylene Graduated Beaker with Closed Handle and 2 Scales have both injection-molded and blue-embossed scales. Graduations are marked in capacity-appropriate increments.
  • Polypropylene beaker
  • Graduated
  • Blue embossed scale
  • Injection molded scale
  • Transparent
Specifications
Accuracy 500 mL
Color Transparent
Diameter (Metric) Outer 90 mm
Capacity (Metric) 500 mL
Material Polypropylene
For Use With (Application) For Use in The Laboratory, Workshop and Household
Height (Metric) 135 mm
Includes With Injection Molded Scale and Blue Embossed Scale
Increments 10 mL
Quantity 1 Ea.
Type Beaker
Autoclavable Autoclavable
Graduated Yes
Scale Blue Graduated Scale
Style Closed Handle