Learn More
Witeg™ Polypropylene Graduated Cylinder
Brand: Witeg™ 7 000 007
Additional Details : Weight : 0.80000kg
Specifications
Polypropylene | |
Embossed | |
6Pack | |
Round | |
439 mm |
6Pack | |
66 mm | |
10 mL | |
Hexagonal | |
Graduated Cylinder |