missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mutexil™ Polypropylene Hooded Combination Coveralls
Click to view available options
Size:
Large
X-Large
2X-Large
Color:
Blue
White
Unit Size:
Pack of 50
Description
- Polypropylene non-woven coverall with hood
- Elastic wrists, ankles, and back
- Zip fastening and two pockets
- For maintenance, production site visits
- Non hazardous environments
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Closure Type | Front Zipper |
| Sterility | Non-sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction