Learn More
Brady™ Polypropylene: ISO Safety Sign - Wear protective clothing
Sign compliant to ISO 7010.
Brand:  Brady™ 819321
Additional Details : Weight : 0.19200kg
 View more versions of this product
Product Code. 15674497
  • 37.80€ / Each
Estimated Shipment:
to see stock.
Description
Description
  • Polypropylene
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
1 Ea.
Indoor and Outdoor
-40°C to 120°C
Global
Polypropylene
400 mm
1.501 mm
Round
White on Blue