missing translation for 'onlineSavingsMsg'
Learn More

Mutexil™ Polypropylene Lab Coats

Product Code. 11790514
Click to view available options
Size:
X-Large
2X-Large
3X-Large
Color:
Blue
White
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11790514

Brand: Mutexil™ 4180171XXXL

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Polypropylene lab coats for use in laboratory settings. Available in a variety of sizes.

PP non woven labcoat with press-stud fastening and pockets
  • Disposable
  • PP, 40g/m2
  • For maintenance, production site visits
  • For non hazardous environments

Specifications

Product Type General Purpose Lab Coat
Size 3X-Large
Gender Unisex
Material Polypropylene
Color White
Cuff Style Elastic
Disposable Single-use
Sterility Sterile
No. of Pockets 2
For Use With (Application) Non-hazardous environments with a high frequency change of clothing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.