Learn More
Masterflex™ Polypropylene Luer Fitting
Product Code. 15114865
Click to view available options
For Use With:
Female Luer x 1/4 to 28 in. UNF
Female Luer x Female Luer, Elbow
Female x Female x Female
Male LL to 1/4 to 28 UNF Thread
Male LL to 10 to 32 Thread
Male x Female x Female
Unit Size:
Pack of 25
Product Code. 15114865
Brand: Masterflex™ 4550866
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Featuring a variety of fitting options, these Luer fittings are ideal for many applications including syringes or small tubing. Cole-Parmer™ Polypropylene Luer Fitting are available with standard, wide, and low-profile barbs.
  • Three barb styles: standard, wide-bore, and low-profile.
    • Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing.
    • Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene.
  • Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable.
  • All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Choose from a range of sizes, 1/16in. to 1/4in., with a variety of connection options.
  • Pack of 25
Specifications
Certifications/Compliance ISO 594-1 standards
Material Polypropylene
For Use With Female Luer x 1/4 to 28 in. UNF
Packaging Quantity 25 Pack
Product Type Luer Fitting