Learn More
Cole-Parmer™ Polypropylene Manifold
Product Code. 11715258
Click to view available options
Outlet Port:
10 Outlets
6 Outlets
Unit Size:
Each
Product Code. 11715258
Brand: Cole-Parmer™ 3152217
Only null left
Add to basket
Add to basket
Expand your flow and line capacity
  • Constructed of durable polypropylene.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Material Polypropylene
Overall Length (Metric) 17.15 cm
Min. Operating Temperature (English) 32°F
Min. Operating Temperature (Metric) 0°C
Tubing Size 1/2 in.
Inlet 1.2 in. NPT(F)
Outlet Port 6 Outlets
Width (Metric) 3.81 cm
Height (Metric) 3.81 cm
Maximum Pressure 150 psi
Max. Operating Temperature (English) 210°F
Max. Operating Temperature (Metric) 99°C
Product Type Manifold