Learn More
Cole-Parmer™ Polypropylene Manifold
Product Code. 11775258
Click to view available options
Outlet Port:
10 Outlets
6 Outlets
Unit Size:
Each
Product Code. 11775258
Brand: Cole-Parmer™ 3152229
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Expand your flow and line capacity
  • Constructed of durable polypropylene.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Material Polypropylene
Overall Length (Metric) 27.31 cm
Min. Operating Temperature (English) 32°F
Min. Operating Temperature (Metric) 0°C
Tubing Size 1/2 in.
Inlet 1.2 in. NPT(F)
Outlet Port 10 Outlets
Width (Metric) 3.81 cm
Height (Metric) 3.81 cm
Maximum Pressure 150 psi
Max. Operating Temperature (English) 210°F
Max. Operating Temperature (Metric) 99°C
Product Type Manifold