Learn More
Fisherbrand™ Polypropylene Microtubes with Snap Cap
Product Code. 11921869
Click to view available options
Color:
Blue
Green
Red
Yellow
Unit Size:
Pack of 1000
Product Code. 11921869
Brand: Fisherbrand™ 47114
Only null left
Add to basket
Edge
Add to basket
Provide a safe, liquid-tight, reliable seal even with prolonged boiling using Fisherbrand™ Polypropylene Microtubes. Use these polypropylene tubes for sample preparation, centrifugation or storage.The microtubes provide convenient, ergonomic and reliable one-handed operation.
  • Flat snap cap
  • Autoclavable
  • Polypropylene
  • Eppendorf style
Specifications
Cap Style Flat cap
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Green
Material Polypropylene
Height (Metric) 40mm
No. per Pack 1000/Pk.
Volume (Metric) 1.5mL
Graduated No
Diameter (Metric) 10 mm