Learn More
Fisherbrand™ Polypropylene Natural Universal Pipet Tip
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 to 200 μL
200 to 1000 μL
Unit Size:
Pack of 120
Pack of 960
Description
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Specifications
Specifications
| Volume (Metric) | 1 to 200 μL |
| Color | Natural |
| Autoclavable | Autoclavable |
| Format | Rack |
| For Use With (Equipment) | MultiTray |
| Material | Polypropylene |