missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Natural Universal Pipet Tip

Product Code. 12369375 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 to 200 μL
200 to 1000 μL
Unit Size:
Pack of 120
Pack of 960

Product Code. 12369375

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Natural Universal Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.

  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types

Specifications

Volume (Metric) 200 to 1000 μL
Color Natural
Autoclavable Autoclavable
For Use With (Equipment) MultiTray
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.