missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Pipet Tip xl

Product Code. 10123673
Click to view available options
Volume (Metric):
0.1 to 10 μL
Unit Size:
Pack of 960
This item is not returnable. View return policy

Product Code. 10123673

Brand: Fisherbrand™ 2530192

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Volume (Metric) 0.1 to 10 μL
Color Crystal
Autoclavable Autoclavable
Length (Metric) 3.83cm
Certifications/Compliance CE, IVD
For Use With (Application) Ideal for All Commercially Available Pippets
Sterilization Method Autoclavable
Format Rack
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.