Learn More
Mutexil™ Polypropylene Protection Sleeves
46.87 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Color:
Transparent
White
Description
- With elastics at both ends
- Length: 40cm
- White, Transparent, or Blue
Specifications
Product Type | General Purpose Sleeve |
Material | Polyethylene |
Color | Transparent |
Disposable | Single-use |
Length (Metric) | 40 mm |
Width (Metric) | 20 mm |