Learn More
Macherey-Nagel™ Polypropylene Reservoir Column
Product Code. 10621502 Shop All Macherey-Nagel Products
Click to view available options
For Use With (Equipment):
1, 3, 6 mL Chromabond Polypropylene Columns
15, 45, 70 mL Chromabond Polypropylene Columns
Quantity:
1 Pack
10 Pack
Unit Size:
Each
Pack of 10
Product Code. 10621502
Brand: Macherey-Nagel™ 730388
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Column Capacity (Metric) 70 mL
Column Type Reservoir
For Use With (Equipment) 15, 45, 70 mL Chromabond Polypropylene Columns
Type Reservoir Column
Includes Adapter
Quantity 1 Pack