Learn More
Azpack™ Polypropylene Round Buckets with Plastic Handle
Product Code. 13139320
Click to view available options
Capacity (Metric):
15 L
Diameter (Metric) Outer:
318 mm
Height (Metric):
292.5 mm
Unit Size:
Each
Product Code. 13139320
Brand: Azpack™ 13139320
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Store, lift, and transport food items and other contents with less effort. Azpack™ Polypropylene Round Buckets with Plastic Handle are also suitable for collecting, carrying and storing biological samples.
  • Constructed of white food grade polypropylene
  • UN approved
  • Supplied with handles and lids
  • Suitable for packaging food
Specifications
Product Type Bucket
Capacity (Metric) 15 L
Material Polypropylene
Color White
Diameter (Metric) Outer 318 mm
Handle Material Plastic
Height (Metric) 292.5 mm
Includes Handle and Lid
Quantity Ea.