missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Polypropylene Screw Caps

Product Code. 10462181
Click to view available options
Color:
Black
Red
Closure Size:
10-425
13-425
8-425
Type:
Large Open Hole Top Screw Cap
Open Top Screw Cap
Screw Cap
Unlined Solid Top Screw Cap
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 10462181

Brand: JG Finneran Associates™ 531013

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Black
Material Polypropylene
Septum Molded
Style WISP style
Quantity 1000 Pk.
Type Open Top Screw Cap
Closure Size 13-425
For Use With (Equipment) Autosamplers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.