missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JG Finneran Associates™ Polypropylene Screw Caps
Click to view available options
Color:
Black
Red
Closure Size:
10-425
13-425
8-425
Type:
Large Open Hole Top Screw Cap
Open Top Screw Cap
Screw Cap
Unlined Solid Top Screw Cap
Unit Size:
Pack of 1000
Specifications
Specifications
| Color | Red |
| Material | Polypropylene |
| Septum | Molded |
| Style | Large Open Hole Top Screw |
| Quantity | 1000 Pk. |
| Type | Screw Cap |
| Closure Size | 8-425 |
| For Use With (Equipment) | Shimadzu with Flange, Dram Vials |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction