Learn More
Atlas Material Testing™ Polypropylene Special Cartridge
Brand: Atlas Material Testing™ RB5115219
Additional Details : Weight : 0.18000kg
Specifications
Food Quality | |
1000 L/hr. | |
1 Each | |
35°C | |
254 mm |
Threshold: 100μM | |
Polypropylene | |
35°C | |
Cartridge |