Learn More
Vitlab™ Polypropylene Spray Bottles
Product Code. 10302611
Change view
Click to view available options
Capacity (Metric):
400 mL
850 mL
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10302611 400 mL Each
12674970 400 mL Pack of 5
10463291 850 mL Each
15252509 850 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10302611 Supplier Vitlab™ Supplier No. 53510
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use these bottles for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography. VITLAB™ Polypropylene Spray Bottles feature a sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle.
  • White polypropylene spray bottles
  • Spray nozzle can be regulated from the finest mist (nebulizing) to a precise liquid jet
  • Range: approx. 3 to 4 meters
Specifications
Capacity (Metric) 400 mL
Color White
For Use With (Application) Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography
Material Polypropylene
No. per Pack 5 Pack
Closure Color White
Closure Included Yes
Product Type Spray Bottle
Closure Material Polypropylene
Closure Type Screw Cap
Autoclavable Not Autoclavable
Show More Show Less
For research purposes only