Learn More
Ismatec™ Standard Tubing Connectors, Plastic
Variously sized and shaped tubing connectors. Ismatec™ Standard Tubing Connectors, Plastic are available in different plastic compositions.
Brand:  Ismatec™ ISM556A
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10007181
  • 16.05€ / Pack of 10
Estimated Shipment: 09-06-2022
to see stock.
Add to basket
Description
Description
  • Standard, non-reducing.
  • Designed for Ismatec™ pump tubing.
  • Available in different sizes, shapes, and compositions.
Specifications
Specifications
Acetal Resin (POM, Polyoxymethylene)
For 1.5mm I.D. Tubing
1.5mm O.D.
Standard Tubing Connector
10/Pk.
Straight