missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Polypropylene Stoppers

Product Code. 30090408
Click to view available options
Size:
NS 19/26
NS 24/29
NS 29/32
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 30090408

Brand: Hirschmann™ 8900275

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A simple and economical choice for the closure of vessels.

  • Manufactured from polypropylene
  • With standard ground
  • Available in a range of sizes

Specifications

For Use With (Application) Closing vessels
Size NS 19/26
Color Natural
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.