missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Polypropylene Stoppers
Click to view available options
Size:
NS 19/26
NS 24/29
NS 29/32
Unit Size:
Each
Pack of 10
Description
- Manufactured from polypropylene
- With standard ground
- Available in a range of sizes
Specifications
Specifications
| For Use With (Application) | Closing vessels |
| Size | NS 19/26 |
| Color | Natural |
| Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction