Learn More
Bel-Art™ SP Scienceware™ Polypropylene Thermometer Joint Adapter
Joint Adapters have a Teflon™ PTFE body with Viton™ O-ring for air tight vacuum work
Brand: Bel-Art™ SP Scienceware™ F20309-1680
Additional Details : Weight : 0.45000kg
Description
- Adapter has a threaded polypropylene screw cap on top with an inside O-ring for depth adjustment of 6-7mm for straight stem thermometers, glass tubing and other items
- Rotating the extracting nut removes the adapter from flask
- Not autoclavable
Specifications
6 to 7 mm | |
Screw cap with inside O-ring | |
Thermometer Joint Adapter |
For 14/20 Tapered Joints | |
Polypropylene, PTFE | |
Straight Stem Thermometers, Glass Tubing |