Learn More
Buerkle™ Polypropylene Thread Adapter
Click to view available options
Thread Size:
External: 2 in. BSP, External 61 mm
External: 2 in. Coarse Tri-Sure - 64 mm (BSI)
External: 64 mm (BSI), Internal: DIN 51
External: 64 mm (BSI), Internal: DIN 60
External: 64 mm (BSI), Internal: DIN 61
External: 64 mm (BSI), Internal: DIN 71
External: Mauser, Internal: 2 in. BSP
External: Tri-Sure, Internal: 2 in. BSP
External: Tri-Sure, Internal: Mauser
External: US Drum Thread, Internal: 2 in. BSP
Internal: 51 mm, 2 in. BSP
Internal: 71 mm, 2 in. BSP
Internal: DIN 61 - 2 in. BSP
Color:
Black
Blue
Blue, Brown
Brown
Green
Orange
Red
Silver
Yellow
Yellow, Blue
Unit Size:
Each
Description
- Constructed of polypropylene
- Interchange the connections to different adapters
Specifications
Specifications
| Material | Polypropylene |
| For Use With | For 0600 Series Model Pumps |
| Tubing Size | DIN 51 |
| Thread Size | External: 64 mm (BSI), Internal: DIN 51 |
| Includes | External Thread 64 mm (BSI), Internal Thread DIN51 |
| Color | Black |
| Product Type | Thread Adapter |