missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL36G / IL1F9 Protein

Product Code. 15953558
Click to view available options
Quantity:
10 μg
100 μg
2 μg
250 μg
Unit Size:
100µg
10µg
250µg
2µg
This item is not returnable. View return policy

Product Code. 15953558

Brand: enQuireBio™ QP10341ec2ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human IL36G / IL1F9, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001265497.1
Gene ID (Entrez) 56300
Name IL36G / IL1F9 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol IL36G
Protein Length Partial (see sequence information for more details).
Biological Activity The specific activity is determined by its binding ability in a functional ELISA. Immobilized rHuIL-36gamma at 1 ug/mL can bind recombinant human IL-1 Rrp2 Fc Chimera with a range of 0.15-5 ug/mL.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MRGTPGDADG GGRAVYQSMC KPITGTINDL NQQVWTLQGQ NLVAVPRSDS VTPVTVAVIT CKYPEALEQG RGDPIYLGIQ NPEMCLYCEK VGEQPTLQLK EQKIMDLYGQ PEPVKPFLFY RAKTGRTSTL ESVAFPDWFI ASSKRDQPII LTSELGKSYN TAFELNIND
Buffer 0.2 um filtered PBS, pH 7.4, lyophilized
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.