Learn More
Semadeni™ Polypropylene Tweezers
Click to view available options
Length (Metric):
125 mm
Unit Size:
Pack of 100
Description
- Autoclavable
- White in color
- 125mm in length
Specifications
Specifications
| Material | Polypropylene |
| Length (Metric) | 125 mm |
| Product Type | Tweezers |
| Quantity (No. per Case) | 100/Pack |
| Type | Tweezers |