Learn More
Masterflex™ Polypropylene Union Compression Connector Compression Fittings
Product Code. 11856284
Click to view available options
For Use With:
0.25 in. tubing O.D.
0.375 in. tubing O.D.
0.5 in. tubing O.D.
0.625 in. tubing O.D.
1/4 in. tubing O.D.
3/8 in. tubing O.D.
Shape:
Elbow
Straight
Unit Size:
Pack of 10
Product Code. 11856284
Brand: Masterflex™ 0638330
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Compression fittings are ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics, and metals.
  • With nut that compresses rigid tubing against fitting body for high-pressure applications
  • Withstands higher pressures than barbed fittings
  • Forms a secure seal - no leaks, flow restrictions or costly downtime
  • Sterilization through ethylene oxide (ETO) only
Specifications
Material Polypropylene
For Use With 3/8 in. tubing O.D.
Min. Operating Temperature (Metric) 0°C
Packaging Quantity 10 Pack
Port Size 1/4 in. orifice; 1-11/32 x 1-11/32 in.
Sterilization Method Ethylene Oxide
Shape Elbow
Maximum Pressure 220 psi
Max. Operating Temperature (Metric) 100°C
Product Type Compression Fitting