Learn More
Fisherbrand™ Yellow Pipet Tip “E”, MultiTray™ 10 x 96
Use these pipette tips are ideal for all commercially available pipets. ratiolab™ Yellow Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets. Available in MultiTray™ 10 x 96 rack.
Brand: Fisherbrand™
Additional Details : Weight : 0.55800kg
Description
These pipette tips offer uniform high quality and precision. Together with specially developed Multirack, Refill System and the ratiolab™ Multibox with the associated pipet tips in Multitray, make pipetting much simpler and safe in the most sensitive applications.- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Specifications
1 to 200 μL | |
Autoclavable | |
CE, IVD | |
Autoclavable | |
Universal Fit |
Yellow | |
4.55cm | |
Ideal for All Commercially Available Pippets | |
Rack | |
Polypropylene |