Learn More
Fisherbrand™ Yellow Pipet Tip “E”, MultiTray™ 10 x 96
Use these pipette tips are ideal for all commercially available pipets. ratiolab™ Yellow Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets. Available in MultiTray™ 10 x 96 rack.
Brand:  Fisherbrand™
Additional Details : Weight : 0.55800kg
Product Code. 10406503
  • 34.80€ / Pack of 960
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
These pipette tips offer uniform high quality and precision. Together with specially developed Multirack, Refill System and the ratiolab™ Multibox with the associated pipet tips in Multitray, make pipetting much simpler and safe in the most sensitive applications.
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Specifications
1 to 200 μL
Autoclavable
CE, IVD
Autoclavable
Universal Fit
Yellow
4.55cm
Ideal for All Commercially Available Pippets
Rack