Learn More
Polysorbate 20, Multi-Compendial, N.F., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
20 mL
200 mL
1 L
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Steel Drum
Unit Size:
1 L
19 L
20mL
4 L
Each
Specifications
Specifications
| Quantity | 4 L |
| Flash Point | 148.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 1.11kg |
| IUPAC Name | 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| Grade | Multi-Compendial/NF |
| Chemical Name or Material | Polysorbate 20 |