Learn More
Greiner Bio-One Polystyrene 384-well Small Volume Microplates
Click to view available options
Color:
Black
Unit Size:
Pack of 40
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Traceable to production through a defined LOT number system
- Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
- Solid bottom
- Well profile: Small Volume™
Specifications
Specifications
| Certifications/Compliance | DIN ISO 9001 |
| Certified | Yes |
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Packaging | Bags of 10 |
| Color | Black |
| Product Type | 384-Well Microplate |
| Material | Polystyrene |
| No. of Wells | 384 |
| Show More |