Learn More
Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes
Used for easy stacking. Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes is designed for better heat resistance using hot agar.
Brand: Greiner Bio-One 633102
16.80 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 8.55000kg
Description
- Vented for better gas exchange
- Heavy design for heat resistance using hot agar
- Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications
Single-use | |
Round | |
480 | |
16 mm |
Polystyrene | |
Non-sterile | |
Yes | |
94 mm |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.