missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Non-Vented Polystyrene Petri Dishes

Used for easy stacking

Brand:  Greiner Bio-One 632102

Additional Details : Weight : 9.90000kg

 View more versions of this product

Product Code. 10567571

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • Heat resistant up to 60°C
  • Available in vented and non-vented
  • Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
  • Easy stacking
  • Crystal polystyrene for optimal analysis by microscopy
Specifications

Specifications

Single-use
Round
Heavy
No
94 mm
Polystyrene
Non-sterile
480 Pack
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.