Learn More
Azpack™ Polystyrene Storage Boxes
Product Code. p-8002489
Click to view available options
Length (Metric):
79 mm
Height (Metric):
22 mm
29 mm
Includes:
Ball-Hinged Lid
Push-Fit Lid
Unit Size:
Pack of 24
Pack of 36
Product Code. 12398278
Brand: Azpack™ 12398278
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
View contents easily with crystal clear construction. Azpack™ Polystyrene Storage Boxes keep laboratory tools and materials organized.
  • Clear polystyrene construction
  • Push-fit, hinged or loose lid included
Specifications
Closure Type Hinged Lid
Color Transparent
Material Polystyrene
Shape Rectangular
For Use With (Application) Storage
Includes Ball-Hinged Lid
Product Type Storage Box
Quantity 24 Pack
Length (Metric) 79 mm
Length (Metric) Interior 73 mm
Width (Metric) 54 mm
Width (Metric) Interior 50 to 51 mm
Height (Metric) 29 mm
Height (Metric) Interior 26 mm