missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinyl Pyrrolidone, Technical, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
Specifications
Specifications
| Color | White |
| pH | 3 to 7 |
| CAS | 9003-39-8 |
| Physical Form | Solid |
| Melting Point | 100°C |
| Molecular Formula | (C6H9NO)n |
| MDL Number | MFCD01076626 |
| Synonym | n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone |
| Quantity | 100 g |
| InChI Key | WHNWPMSKXPGLAX-UHFFFAOYSA-N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction