Learn More
RETSCH POM Dividing Head
Ensure the representativeness of a sample and the reproducibility of the analysis with this dividing head. Retsch™ POM Dividing Heads divide the sample so exactly that the composition of each fraction of the sample corresponds exactly to that of the original bulk sample.
Brand:  RETSCH 42.793.0007
Additional Details : Weight : 3.50000kg
Product Code. 10523542
  • 3290.00€ / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
  • For use with Retsch™ PT 100 Sample Divider
Specifications
Specifications
Dividing Head
Retsch™ PT 100 Sample Divider
POM