Learn More
Rogo-Sampaic Porcelain Buchner Funnels
Click to view available options
Capacity (Metric):
40 mL
80 mL
150 mL
265 mL
490 mL
660 mL
1080 mL
2000 mL
4200 mL
10 L
Diameter (Metric) Plate:
45mm
55mm
70mm
90mm
110mm
125mm
150mm
185mm
240mm
320mm
Unit Size:
Each
Description
- White porcelain
- Available for a range of volumes and filter diameters
- Applications include collecting recrystalized compounds
- Excellent thermal and chemical resistance
Specifications
Specifications
| Capacity (Metric) | 10 L |
| Material | Porcelain |
| Diameter (Metric) Plate | 320mm |
| Diameter (Metric) Disc | 320mm |
| Length (Metric) Stem | 133mm |
| Quantity | 1Each |
| Product Type | Buchner Funnel |