Learn More
Haldenwanger™ Porcelain Gooch Crucibles
Product Code. 12045103
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Each
Product Code. 12045103
Brand: Haldenwanger™ L 82A/3
Only null left
Add to basket
Add to basket
Order Info
DiameterMetricOuter
Specifications
Material Porcelain
Capacity (Metric) 25 mL
Diameter (Metric) Outer 42mm
Height (Metric) 45mm
Product Type Gooch crucible