Learn More
Porcelaines Avignon™ High Form Porcelain Crucibles
Heat and cool substances in the laboratory using Porcelaines Avignon™ Porcelain High Form Crucibles. They are available in various capacities.
Brand:  Porcelaines Avignon™ 11205
Additional Details : Weight : 0.16000kg
 View more versions of this product
Product Code. 11718483
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • High walls
  • Withstand high temperatures and rapid temperature changes
  • Porcelain
Specifications
Specifications
Porcelain
40mm
Crucible
30 mL
45mm