Learn More
Porcelaines Avignon™ High Form Porcelain Crucibles
Heat and cool substances in the laboratory using Porcelaines Avignon™ Porcelain High Form Crucibles. They are available in various capacities.
Brand: Porcelaines Avignon™ 11207
Additional Details : Weight : 0.39600kg
Description
- High walls
- Withstand high temperatures and rapid temperature changes
- Porcelain
Specifications
Porcelain | |
55mm | |
Crucible |
80 mL | |
63mm |