Learn More
Haldenwanger™ Porcelain Medium High Form Melting Crucible
Product Code. 12343143
Click to view available options
Capacity (Metric):
38 mL
Unit Size:
Each
Product Code. 12343143
Brand: Haldenwanger™ 8250145
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Order Info
CapacityMetric
Specifications
Material Porcelain
Capacity (Metric) 38 mL
Diameter (Metric) Outer 45mm
Height (Metric) 36mm
Product Type Melting Crucible