missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heinz Herenz High-Density Polyethylene Waste Bags
Click to view available options
Capacity (English):
1.32 gal.
5.81 gal.
7.04 gal.
19.02 gal.
Unit Size:
Each
Description
- Waste bags
- Yellow for easy identification
- For the disposal of contaminated labware
- Rectangle white indicator patch field turns into dark when autoclaved
- With BIOHAZARD symbol and safety imprint in English, German, French, and Spanish languages
- Autoclavable at 121 °C for 20 minutes (the bag must not be completely closed)
- Thickness 50 μm (0.05 mm)
- HD-PE
Specifications
Specifications
| Capacity (Metric) | 72 L |
| Capacity (English) | 19.02 gal. |
| Color | Yellow |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Material | HDPE |
| Product Type | Biohazard Waste Bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction