missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ KappaXL Affinity HPLC Columns

Product Code. 18436628 Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions:
2.1 x 30 mm
4.6 x 100 mm
4.6 x 50 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions unitSize
18436628 2.1 x 30 mm Each
18476618 4.6 x 50 mm Each
18496438 4.6 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18436628 Supplier Thermo Scientific™ Supplier No. undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ POROS™ CaptureSelect™ KappaXL Affinity HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect affinity ligands.

POROS CaptureSelect KappaXL Affinity HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect™ affinity ligands.

CaptureSelect KappaXL Affinity HPLC columns recognize the Kappa light chain of human immumoglobulin, enabling quantitation and small-scale purification of all immunoglobulins (including IgA, IgM, IgD, IgE), Fab fragments, and bi-specific antibodies containing the Kappa light chain.

Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

Features of POROS CaptureSelect KappaXL Affinity HPLC columns include:

  • Versitile—applicable for human immunoglobulins, Fab fragments, and bi-specific antibodies containing a Kappa light chain
  • Rapid—POROS resins enable quantitation or purification of immunoglobulins and antibodies in minutes
  • Hands-off operation—HPLC method can be automated for a streamlined operation
  • Robust—designed to deliver high-performance purification over hundreds of chromatography cycles

High speed with high specificity

  • POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis, including mass spectrometry.

Automation

POROS Affinity HPLC columns are for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligand enables one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regimen results in strong re-use performance. POROS Affinity HPLC columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

Main characteristics

  • Protein binding capacity: 16.5 mg/mL
  • Species specificity: human
  • Column material: PEEK polymer
  • Frit: 2 μm, titanium
  • POROS resin: cross-linked poly(styrene-divinylbenzene)
  • CaptureSelect ligands: single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae

Specifications

Column Format Analytical Column, Biocompatible PEEK
Diameter (Metric) 2.1 mm
Length (Metric) 30 mm
Max. Pressure 2400 psi (170 bar)
Particle Size 20 μm
pH 1-10
Pore Size 2000 Å
Quantity 1 Column
Temperature 2-40°C
Stationary Phase Light Chain Kappa affinity ligand, human‐specific
Content And Storage Column includes EZ grip fittings and column test certificate.

Store at 2°C to 8°C. DO NOT FREEZE.

Product Line POROS CaptureSelect
Binding Capacity 16.5 mg/ml
Shipping Condition Wet Ice
Dimensions 2.1 x 30 mm
Packing Material Highly Crosslinked Styrene-Divinylbenzene polymer (ST-DVB)
Column Type Affinity
Material PEEK
Technique Bio-LC
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.